Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Thermo Scientific Chemicals

Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)

Maximum level of impurities: Insoluble matter 0.01%, Cl 0.001%, PO4 5ppm, SO4 0.003%, NH4 5ppm, Ca 0.002%, Mg 0.001%, Heavy Metals (as Pb) 5ppm, Fe 5ppm, Na 0.03% | CAS: 298-14-6 | CHKO3 | 100.11 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 040195.36
亦稱為 040195-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
檢視更多
規格Specification Sheet規格表
Assay (unspecified)99.7-100.5% KHCO₃
Impurity contentSO₄ 0.003%
Trace Metals(as Pb) 5ppm
Impurity contentCa 0.002%
Impurity contentCl 0.001%
檢視更多
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only