Thermo Scientific Chemicals

Sodium iodide, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 040196.22
100 g, Each
Thermo Scientific Chemicals

Sodium iodide, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 040196.22
100 g, Each
Quantity
Catalog number: 040196.22
also known as 040196-22
Price (USD)
Price: 76.10
Online price: 64.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7681-82-5
IUPAC Name
sodium iodide
Molecular Formula
INa
InChI Key
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES
[Na+].[I-]
Assay from Suppliers CofA
≥99.5%
Impurity content
Barium, Ba: ≤0.002%
Impurity content
Chloride, Cl and Bromide, Br (as Cl): ≤0.01%
Impurity content
Phosphate, PO₄: ≤0.001%
Trace Metals
Heavy metals as Lead, Pb: ≤5ppm

Description

Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
  • Commonly used to prevent iodine deficiency Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines
  • Used in baking ingredient conditioner and as animal feed
  • Used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text