Thermo Scientific Chemicals

Ammonium iodide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 040255.14
25 g, Each
Thermo Scientific Chemicals

Ammonium iodide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 040255.14
25 g, Each
Quantity
Catalog number: 040255.14
also known as 040255-14
Price (USD)
Price: 253.00
Online price: 215.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12027-06-4
IUPAC Name
ammonium iodide
Molecular Formula
H4IN
InChI Key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES
[NH4+].[I-]
Appearance (Color)
White
Assay (special)
Iodine: 85.5-89.5%
Total Metal Impurities
≤0.005% (50 ppm)
Form
Crystals

Description

It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text