Europium(III) trifluoromethanesulfonate, 98%
Europium(III) trifluoromethanesulfonate, 98%
Europium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Europium(III) trifluoromethanesulfonate, 98%

CAS: 52093-25-1 | C3EuF9O9S3 | 599.15 g/mol
Have Questions?
產品號碼Quantity
040305.09
亦稱為 040305-09
10 g
產品號碼 040305.09
亦稱為 040305-09
價格 (TWD)
-
Quantity:
10 g
Request bulk or custom format
化學識別
CAS52093-25-1
IUPAC Nameeuropium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3EuF9O9S3
InChI KeyTWNOVENTEPVGEJ-UHFFFAOYSA-K
SMILES[Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
檢視更多
規格Specification Sheet規格表
Assay (unspecified)97.6% min
It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only