Thermo Scientific Chemicals

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Catalog number: 040342.30
250 g, Each
Thermo Scientific Chemicals

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Catalog number: 040342.30
250 g, Each
Quantity
Catalog number: 040342.30
also known as 040342-30
Price (USD)

Chemical Identifiers

CAS
7446-18-6
IUPAC Name
bis(λ¹-thallanylium) sulfate
Molecular Formula
O4STl2
InChI Key
YTQVHRVITVLIRD-UHFFFAOYSA-L
SMILES
[Tl+].[Tl+].[O-]S([O-])(=O)=O
Total Metal Impurities
0.5% max.

Description

Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text