Thermo Scientific Chemicals

Aluminum oxide, Aerosol Refractory Paint, Thermo Scientific Chemicals

Catalog number: 040391.0CL
5 x 13 oz., Each
Thermo Scientific Chemicals

Aluminum oxide, Aerosol Refractory Paint, Thermo Scientific Chemicals

Catalog number: 040391.0CL
5 x 13 oz., Each
Quantity
Catalog number: 040391.0CL
also known as 040391-0:
Price (USD)
Price: 856.00
Online price: 740.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Comment
Maximum temperature use: 1800?C Air, vacuum or inert; 1500?C Vacuum and carbon

Description

Provides easy application of uniform, thin, protective high temperature Aluminum oxide coatings. Substrates can be ceramic, graphite or metal. Aluminum oxide is also widely used as an abrasive. In the paper-making process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Provides easy application of uniform, thin, protective high temperature Al2O3 coatings
  • Substrates can be ceramic, graphite or metal
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text