Buffer solution, pH 1.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Buffer solution, pH 1.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
For Calibration of pH Meters
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
Catalog number 040446.K7
also known as 040446-K7
Price (USD)/ Each
78.60
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
78.60
Add to cart
Buffer solution, pH 1.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Catalog number040446.K7
Price (USD)/ Each
78.60
-
Add to cart
Chemical Identifiers
CAS7447-40-7, 7647-01-0, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofApH 1.00 (± 0.026 @ 25°C)
FormLiquid
Calibration of pH meters

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only