Iron rod, 5mm (0.2in) dia, 99.99% (metals basis)
Iron rod, 5mm (0.2in) dia, 99.99% (metals basis)
Iron rod, 5mm (0.2in) dia, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron rod, 5mm (0.2in) dia, 99.99% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
150 mm
Catalog number 040500.DO
also known as 040500-DO
Price (EUR)
413,65
Online Exclusive
459,00
Save 45,35 (10%)
Each
Quantity:
150 mm
Request bulk or custom format
Price (EUR)
413,65
Online Exclusive
459,00
Save 45,35 (10%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 5mm (0.2in) diameter.
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only