Titanium(IV) iodide, anhydrous, 99.9% (metals basis)
Titanium(IV) iodide, anhydrous, 99.9% (metals basis)
Titanium(IV) iodide, anhydrous, 99.9% (metals basis)
Thermo Scientific Chemicals

Titanium(IV) iodide, anhydrous, 99.9% (metals basis)

CAS: 7720-83-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 040663.09
also known as 040663-09
Price (EUR)
221,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
221,00
Each
Chemical Identifiers
CAS7720-83-4
IUPAC Nametitanium(4+) tetraiodide
Molecular FormulaI4Ti
InChI KeyNLLZTRMHNHVXJJ-UHFFFAOYSA-J
SMILES[Ti+4].[I-].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Titanium(IV) iodide, anhydrous is used in the CVD of titanium nitride films with optimized characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only