Germanium n-butoxide, 97+%
Germanium n-butoxide, 97+%
Germanium n-butoxide, 97+%
Thermo Scientific Chemicals

Germanium n-butoxide, 97+%

Oxide films | CAS: 25063-27-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 040816.14
also known as 040816-14
Price (USD)
569.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
569.00
Each
Chemical Identifiers
CAS25063-27-8
Molecular FormulaC16H32GeO4
InChI KeyMPVCRVXMJOZDGL-UHFFFAOYSA-N
SMILES[Ge].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-]
Molecular Weight (g/mol)361.06
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay from Supplier's CofA≥97%
Appearance (Color)Clear colorless
FormLiquid
Germanium n-butoxide is used as oxide films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only