Molybdenum sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Molybdenum sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Molybdenum sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Catalog NumberQuantity
040868.KS1 Ea.
Catalog number 040868.KS
Price (TWD)
-
Quantity:
1 Ea.
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target,76.2mm (3.0in) diameter x 3.18mm (0.125in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only