Thermo Scientific Chemicals

Hastelloy C|r gauze, 100 mesh woven from 0.102mm (0.004in) dia wire, Thermo Scientific Chemicals

Catalog number: 040940.FL
75 x 75 mm, Each
Thermo Scientific Chemicals

Hastelloy C|r gauze, 100 mesh woven from 0.102mm (0.004in) dia wire, Thermo Scientific Chemicals

Catalog number: 040940.FL
75 x 75 mm, Each
Quantity
Catalog number: 040940.FL
also known as 040940-FL
Price (USD)
Price: 52.20
Online price: 44.65
Your price:
Quantity
-
Assay from Suppliers CofA
Co: ≤2.5%
Assay from Suppliers CofA
Cr: 20.0-22.5%
Assay from Suppliers CofA
Fe: 2.0-6.0%
Assay from Suppliers CofA
Mn: ≤0.5%
Assay from Suppliers CofA
Mo: 12.5-14.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text