Thermo Scientific™

Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316, Thermo Scientific™

300 x 300 mm
Thermo Scientific™

Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316, Thermo Scientific™

300 x 300 mm
Quantity
Catalog number: 040943.HB
also known as 040943-HB
Price (USD)
296.00 / -
Your Price:
Quantity
-
Assay (unspecified)
Cr 16.0 - 18.0%
Assay (unspecified)
P 0.045% max
Appearance (Color)
Gray
Transition Temperature
Ni 10.0 - 14.0%
Assay (unspecified)
Mn 2.0% max

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures