Thermo Scientific Chemicals

Methanol, Environmental Grade, >99.8%, Thermo Scientific Chemicals

1 L, Each
Thermo Scientific Chemicals

Methanol, Environmental Grade, >99.8%, Thermo Scientific Chemicals

1 L, Each
Quantity
Catalog number: 040980.K2
also known as 040980-K2
Price (USD)
Price: 43.20
Your Price:
Quantity
-

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Residue on Evaporation
2ppm
Residual halide
5 ng/l (GC-ECD)

Description

It is suitable for environmental and GC analysis. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. Used for producing bio-diesel, synthesis of formaldehyde and acetic acid, useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin. It is useful for the preparation of compact direct methanol fuel cells for portable application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only