Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis), Thermo Scientific Chemicals - FAQs
Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis), Thermo Scientific Chemicals - FAQs
View additional product information for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis), Thermo Scientific Chemicals - FAQs (041000.18, 041000.A7, 041000.A1, 041000.30)
5 product FAQs found
What is the tap density of Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis)?
The tap density of Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis) is 1.4.
What is the iron wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis)?
The iron wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis) is 0.2?
What is the oil and grease wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis)?
The oil and grease wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis) is 0.2.
What is the volatile wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis)?
The volatile wt% maximum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis) is 0.1.
What is the free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis)?
The free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis) is 97.5.