Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis), Thermo Scientific Chemicals - FAQs
View additional product information for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis), Thermo Scientific Chemicals - FAQs (041001.30, 041001.A7, 041001.A1)
5 product FAQs found
What is the tap density of Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?
The tap density of Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 1.4.
What is the iron wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?
The iron wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.2?
What is the oil and grease wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?
The oil and grease wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.2.
What is the volatile wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?
The volatile wt% maximum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 0.1.
What is the free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis)?
The free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 10.0-14.0 micron, 98% (metals basis) is 97.5.