Silicon(IV) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)
Thermo Scientific Chemicals

Silicon(IV) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)

 
Have Questions?
Catalog NumberQuantity
041097.KS1 Ea.
Catalog number 041097.KS
Price (EUR)
452,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
452,00
Each
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialSilicon(IV) oxide sputtering target
EINECS Number231-545-4
Formula Weight60.09
MDL NumberMFCD00011232
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only