Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%
Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%
Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%
Thermo Scientific Chemicals

Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%

CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 041181.MD
also known as 041181-MD
Price (EUR)
97,60
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
97,60
Each
Specifications
CAS99685-96-8
ChEBICHEBI:33128
Chemical Name or MaterialFullerene powder
ColorBlack-Brown
Concentration or Composition (by Analyte or Components)typically 73% C60, 22% C70, higher 5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only