Bismuth(III) tert-pentyloxide, 97+%
Thermo Scientific Chemicals

Bismuth(III) tert-pentyloxide, 97+%

CAS: 124687-44-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041203.03
also known as 041203-03
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.00
Save 11.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.00
Save 11.35 (15%)
Each
Specifications
CAS124687-44-1
Chemical Name or MaterialBismuth(III) tert-pentyloxide
ColorGray
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Formula Weight470.41
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only