Search
View additional product information for Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight - FAQs (041238.14, 041238.36, 041238.22)
1 product FAQs found
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, has an approximate molecular weight of 10,000 g/mol.