Benzalkonium chloride
Benzalkonium chloride
Benzalkonium chloride
Thermo Scientific Chemicals

Benzalkonium chloride

Cationic surface active agent | CAS: 63449-41-2 | C21H38ClN | 339.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 041339.14
also known as 041339-14
Price (EUR)
35,90
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
35,90
Each
Chemical Identifiers
CAS63449-41-2
IUPAC Namebenzyl(dodecyl)dimethylazanium chloride
Molecular FormulaC21H38ClN
InChI KeyJBIROUFYLSSYDX-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormSolid to semi-solid
Assay (unspecified)95 to 105% ( dry basis)
Cationic surface active agentBenzalkonium chloride is used in textile dyeing, softeners, antistatic agents, emulsifiers and conditioner. It acts as a cationic detergent. It is also used as a preservative in pharmaceutical products like eye, ear and nasal drops and sprays. Further, it is an active ingredient in personal care products such as hand sanitizers, shampoos, deodorants and wet wipes. In addition to this, it is used in a study to assess its action on epithelia conjunctival cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only