Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Thermo Scientific Chemicals

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)

Precursor for CVD process | CAS: 13986-53-3 | C22H38CoO4 | 425.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 041359.04
also known as 041359-04
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS13986-53-3
Chemical Name or MaterialBis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)
Formula Weight425.46
Health Hazard 1H317-H341
View more
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only