Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis)
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis)
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis)
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis)
Thermo Scientific Chemicals

Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 041370.36
also known as 041370-36
Price (USD)
1,088.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,344.00
Save 255.35 (19%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
1,088.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,344.00
Save 255.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
FormGranules
Total Metal Impurities≤0.001%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only