Chloroform-d, 100% (Isotopic), contains 0.03% v/v TMS
Chloroform-d, 100% (Isotopic), contains 0.03% v/v TMS
Chloroform-d, 100% (Isotopic), contains 0.03% v/v TMS
Thermo Scientific Chemicals

Chloroform-d, 100% (Isotopic), contains 0.03% v/v TMS

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 Ea.
5 Ea.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041388.KV
also known as 041388-KV
Price (EUR)
-
Quantity:
10 Ea.
Request bulk or custom format
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)>/= 99.5% as Pass/ Fail
Water Content (Karl Fischer Titration)100 ppm, max
Proton NMR>/= 99.96% isotopic enrichment
Concentration v/v0.024 - 0.036 % TMS
View more
Widely used in the organic solvent for the NMR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only