Potassium tert-pentyloxide, 14-18% w/v in cyclohexane
Potassium tert-pentyloxide, 14-18% w/v in cyclohexane
Potassium tert-pentyloxide, 14-18% w/v in cyclohexane
Thermo Scientific Chemicals

Potassium tert-pentyloxide, 14-18% w/v in cyclohexane

Coreactant for conversion of ketones to oxides | CAS: 41233-93-6 | C5H11KO | 126.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041394.14
also known as 041394-14
Price (EUR)
139,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
139,00
Each
Chemical Identifiers
CAS41233-93-6
IUPAC Namepotassium 2-methylbutan-2-olate
Molecular FormulaC5H11KO
InChI KeyZRLVQFQTCMUIRM-UHFFFAOYSA-N
SMILES[K+].CCC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Free base (titration)≤0.20% (as KOH)
Concentration (weight/weight)14.0-16.0%
Appearance (Color)Clear colorless to pale yellow to pale orange
Potassium tert-pentyloxide is used in wide range of applications in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It is also used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only