Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
200 x 200 mm
Catalog number 041451.GH
also known as 041451-GH
Price (EUR)
-
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver foil
EINECS Number231-131-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only