1-(tert-Butyldimethylsilyl)imidazole, 97%
1-(tert-Butyldimethylsilyl)imidazole, 97%
1-(tert-Butyldimethylsilyl)imidazole, 97%
Thermo Scientific Chemicals

1-(tert-Butyldimethylsilyl)imidazole, 97%

CAS: 54925-64-3 | C9H18N2Si | 182.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 041462.03
also known as 041462-03
Price (EUR)
58,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
58,60
Each
Specifications
CAS54925-64-3
Chemical Name or Material1-(tert-Butyldimethylsilyl)imidazole
Density0.94 g/mL
EINECS Number259-398-1
Flash Point99°C (210°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only