Copper(II) ethylacetoacetate, 97%
Copper(II) ethylacetoacetate, 97%
Copper(II) ethylacetoacetate, 97%
Thermo Scientific Chemicals

Copper(II) ethylacetoacetate, 97%

CAS: 14284-06-1 | C12H18CuO6 | 321.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 041489.04
also known as 041489-04
Price (EUR)
35,30
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
35,30
Each
Chemical Identifiers
CAS14284-06-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to blue-green to blue
FormPowder
Assay from Suppliers CofA≥96%
Copper(II) ethylacetoacetate it acts as a common cu(+2) precursor. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only