Thermo Scientific Chemicals

Sulfanilic acid, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 041540.30
250 g, Each
Thermo Scientific Chemicals

Sulfanilic acid, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 041540.30
250 g, Each
Quantity
Catalog number: 041540.30
also known as 041540-30
Price (USD)
Price: 139.00
Online price: 119.65
Your price:
Quantity
-

Chemical Identifiers

CAS
121-57-3
IUPAC Name
4-aminobenzene-1-sulfonic acid
Molecular Formula
C6H7NO3S
InChI Key
HVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)S(O)(=O)=O
Impurity content
SO₄ 0.01%
Impurity content
Insoluble in sodium carbonate 0.02%
Impurity content
NO₄ 0.5ppm
Residue on ignition
0.01%
Impurity content
Cl 0.002%

Description

Sulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text