Polyethylene glycol monomethylether, 550
Polyethylene glycol monomethylether, 550
Polyethylene glycol monomethylether, 550
Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 550

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 041561.22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
規格Specification Sheet規格表
Residual water0.5% max
FormLow melting point solid, or liquid
pH4.5 to 7.0 ( 5% aq sol at 25°C)
Molecular Weight525 - 575 D
Polyethylene glycol monomethylether, 550 is used for formulating screens and for optimization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only