Thermo Scientific Chemicals

Stainless Steel foil, 0.05mm (0.002in) thick, Type 304, Thermo Scientific Chemicals

Catalog number: 041582.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Stainless Steel foil, 0.05mm (0.002in) thick, Type 304, Thermo Scientific Chemicals

Catalog number: 041582.FI
50 x 50 mm, Each
Quantity
Catalog number: 041582.FI
also known as 041582-FI
Price (USD)
Price: 37.80
Online price: 32.65
Your price:
Quantity
-
Appearance (Color)
Silver
Assay (unspecified)
C 0.08% max.
Assay (unspecified)
Cr 18.00 - 20.00%
Assay (unspecified)
Mn 2.00% max.
Assay (unspecified)
N 0.10% max.

Description

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text