Stainless Steel foil, 0.5mm (0.02in) thick, Type 304
Thermo Scientific Chemicals

Stainless Steel foil, 0.5mm (0.02in) thick, Type 304

Fe:Cr:Ni; 70:19:11 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 041585.GH
also known as 041585-GH
Price (EUR)
67,65
Online Exclusive
75,10
Save 7,45 (10%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (EUR)
67,65
Online Exclusive
75,10
Save 7,45 (10%)
Each
Specifications
AppearanceSilver -colored foil
Chemical Name or MaterialStainless Steel
ColorSilver
MDL NumberMFCD00148499
Molecular FormulaFe:Cr:Ni; 70:19:11 wt%
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only