Hafnium ethoxide, 99.9% (metals basis)
Hafnium ethoxide, 99.9% (metals basis)
Hafnium ethoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Hafnium ethoxide, 99.9% (metals basis)

Preparation of refractory coatings | CAS: 13428-80-3 | C8H24HfO4 | 362.766 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041607.06
also known as 041607-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13428-80-3
IUPAC Namehafnium(4+) tetraethanolate
Molecular FormulaC8H20HfO4
InChI KeyBFIMXCBKRLYJQO-UHFFFAOYSA-N
SMILES[Hf+4].CC[O-].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1%
FormPowder or crystals or crystalline powder

Hafnium ethoxide is a chemical used in the preparation of refractory coatings. Some of the other applications include Nano-materials & Thin Films, Display & Imaging, Catalysis & Synthesis, Hybrid & Disperse Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only