Copper(II) methacrylate, tech.
Copper(II) methacrylate, tech.
Copper(II) methacrylate, tech.
Thermo Scientific Chemicals

Copper(II) methacrylate, tech.

CAS: 19662-59-0 | C4H5CuO2 | 148.63 g/mol
製品番号(カタログ番号) 041621.06
または、製品番号041621-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19662-59-0
IUPAC Namecopper(2+) 2-methylprop-2-enoate
Molecular FormulaC4H5CuO2
InChI KeyBFHDHRZKYIRNRP-UHFFFAOYSA-M
SMILES[Cu++].CC(=C)C([O-])=O
さらに表示
Appearance (Color)Blue to green
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only