Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni <0.15%
Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni &lt;0.15%
Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni &lt;0.15%
Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni &lt;0.15%
Thermo Scientific Chemicals

Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni <0.15%

CAS: 12017-01-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 041670.18
also known as 041670-18
Price (EUR)
112,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
112,00
Each
Add to cart
Chemical Identifiers
CAS12017-01-5
SpecificationsSpecification SheetSpecification Sheet
Impurity contentNi < 0.15%
Total Metal Impurities0.2% max.
Cobalt titanium oxide shows magnetic properties at room temperature and used in electronics and as spintronic devices. It is used to improve the gas sensing ability of hydrogen sulfide by hydrothermal method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only