Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh
Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh
Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh
Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh

For the preparation of functionalized beads used in solid-phase synthesis | CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041800.14
also known as 041800-14
Price (EUR)
120,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
120,00
Each
Specifications
CAS9003-70-7
Chemical Name or MaterialPoly(styrene-divinylbenzene)
IUPAC Name1,2-bis(ethenyl)benzene;styrene
InChI KeyBCIPGSZQUDLGSY-UHFFFAOYNA-N
MDL Number9003-70-7
View more
Poly(styrene-divinylbenzene) is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions. is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only