tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
4 x 1 L
Catalog number 041857.M1
also known as 041857-M1
Price (EUR)
525,00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (EUR)
525,00
Each
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay from Supplier's CofA≥99.5%
Residue on Evaporation≤5 ppm
Water Content≤50 ppm
tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only