Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyimide film, 0.008mm thick, Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
100 x 100 mm
150 x 150 mm
300 x 300 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 041896.GH
價格 (TWD)
-
Quantity:
100 x 100 mm
Request bulk or custom format
規格
CAS62929-02-6
Chemical Name or MaterialPolyimide film
IUPAC Name1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
InChI KeyCDTDIQLZIBORMV-UHFFFAOYNA-N
MDL NumberMFCD00677744
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only