Polystyrene latex microsphere, 2 micron, 10 wt% dispersion in water
Polystyrene latex microsphere, 2 micron, 10 wt% dispersion in water
Polystyrene latex microsphere, 2 micron, 10 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 2 micron, 10 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
041918.7C
also known as 041918-7C
15 mL
Catalog number 041918.7C
also known as 041918-7C
Price (USD)
718.65
Online Exclusive
798.00
Save 79.35 (10%)
Each
Quantity:
15 mL
Request bulk or custom format
Price (USD)
718.65
Online Exclusive
798.00
Save 79.35 (10%)
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
FormAqueous suspension
Particle Size2µm ± 0.01
Solid content10% ± 0.5% by weight
Water Content90% ± 0.5% by weight
Use in protein binding; size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only