Aluminum acrylate
Aluminum acrylate
Aluminum acrylate
Thermo Scientific Chemicals

Aluminum acrylate

CAS: 15743-20-1 | C9H12AlO6 | 243.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042003.06
also known as 042003-06
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Specifications
CAS15743-20-1
Chemical Name or MaterialAluminum acrylate
EINECS Number239-836-8
Formula Weight243.17
Health Hazard 1H315-H319-H335
View more
Aluminum acrylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only