Specifications
Chemical Name or MaterialNickel plating solution
ColorGreen
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.
Health Hazard 1H302+H332-H315-H317-H319-H334-H341-H350i-H360FD-H372
Health Hazard 2GHS H Statement
H334-H350-H360-H372-H341-H302-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
View more