Thermo Scientific Chemicals

Diphenyl methylphenyl phosphate, mixture of isomers, 94%, Thermo Scientific Chemicals

Catalog number: 042092.AE
100 mL, Each
Thermo Scientific Chemicals

Diphenyl methylphenyl phosphate, mixture of isomers, 94%, Thermo Scientific Chemicals

Catalog number: 042092.AE
100 mL, Each
Quantity
Catalog number: 042092.AE
also known as 042092-AE
Price (USD)
Price: 44.20
Online price: 37.65
Your price:
Quantity
-

Chemical Identifiers

CAS
26444-49-5
IUPAC Name
4-methylphenyl diphenyl phosphate
Molecular Formula
C19H17O4P
InChI Key
OJUZRFGUKHQNJX-UHFFFAOYSA-N
SMILES
CC1=CC=C(OP(=O)(OC2=CC=CC=C2)OC2=CC=CC=C2)C=C1
Form
Liquid
Refractive Index
1.5580-1.5640 @ 25°C
Appearance (Color)
Clear colorless to pale yellow or pale orange
Assay from Supplier's CofA
≥92.0% (GC) (Sum of isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text