Chromium(III) chloride hexahydrate, 99.5% min
Chromium(III) chloride hexahydrate, 99.5% min
Chromium(III) chloride hexahydrate, 99.5% min
Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, 99.5% min

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 042114.22
亦稱為 042114-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS10060-12-5
IUPAC Namechromium(3+) hexahydrate trichloride
Molecular FormulaCl3CrH12O6
InChI KeyLJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
檢視更多
規格Specification Sheet規格表
Assay (unspecified)99.5% min
Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only