Calcium fluoride crystal optic disc, 38mm x 6mm, polished both sides
Thermo Scientific Chemicals

Calcium fluoride crystal optic disc, 38mm x 6mm, polished both sides

 
Have Questions?
Catalog NumberQuantity
042145.KT1 Ea.
Catalog number 042145.KT
Price (EUR)
318,00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (EUR)
318,00
Each
Specifications
Chemical Name or MaterialCalcium fluoride crystal optic disc
EINECS Number232-188-7
MDL NumberMFCD00010907
Name Note38mm x 6mm, Polished both sides
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only