Thermo Scientific™

Nickel Chromium wire, 0.25mm dia., Thermo Scientific™

100 m
Thermo Scientific™

Nickel Chromium wire, 0.25mm dia., Thermo Scientific™

100 m
Quantity
Catalog number: 042195.H4
also known as 042195-H4
Price (USD)
66.40 / -
Your Price:
Quantity
-

Chemical Identifiers

CAS
11106-97-1
Comment
Ni 80% min
Comment
0.25 mm diameter

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures