Thermo Scientific Chemicals

Silver perchlorate, anhydrous, Thermo Scientific Chemicals

Catalog number: 042209.22
100 g, Each
Thermo Scientific Chemicals

Silver perchlorate, anhydrous, Thermo Scientific Chemicals

Catalog number: 042209.22
100 g, Each
Quantity
Catalog number: 042209.22
also known as 042209-22
Price (USD)
Price: 825.00
Online price: 701.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-93-9
IUPAC Name
silver(1+) perchlorate
Molecular Formula
AgClO4
InChI Key
YDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES
[Ag+].[O-][Cl](=O)(=O)=O
Assay (unspecified)
97% min

Description

Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text