Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042256.30
also known as 042256-30
Price (USD)
708.65
Special offer
Online exclusive
Ends: 31-Dec-2025
833.00
Save 124.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
708.65
Special offer
Online exclusive
Ends: 31-Dec-2025
833.00
Save 124.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
Assay from Supplier's CofA≥99.99% (metals basis)
FormSlug, 3.175mm (0.125in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only