Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042286.KV
10 Ea., Each
Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042286.KV
10 Ea., Each
Quantity
Catalog number: 042286.KV
also known as 042286-KV
Price (USD)

Chemical Identifiers

CAS
2206-27-1
IUPAC Name
(²H₃)methanesulfinyl(²H₃)methane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Appearance (Color)
Colorless
Assay by NMR
99.96% min Isotopic Enrichment
Form
clear liquid

Description

DMSO is a common polar solvent used in NMR, PCR, as a cryoprotectant, and as a universal solvent for many compounds. Deuterated DMSO is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text