Acetonitrile, anhydrous, 99.8+%
Acetonitrile, anhydrous, 99.8+%
Acetonitrile, anhydrous, 99.8+%
Thermo Scientific Chemicals

Acetonitrile, anhydrous, 99.8+%

Water <100ppm, typically 50ppm (0.005%) | CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 042311.AK
also known as 042311-AK
Price (EUR)
49,65
Online Exclusive
55,50
Save 5,85 (11%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
49,65
Online Exclusive
55,50
Save 5,85 (11%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water Content≤100ppm
Assay (unspecified)≥99.8%
Residue on Evaporation≤5ppm
FormLiquid
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only