Poly(2-ethyl-2-oxazoline), M.W. 50,000
Poly(2-ethyl-2-oxazoline), M.W. 50,000
Poly(2-ethyl-2-oxazoline), M.W. 50,000
Thermo Scientific Chemicals

Poly(2-ethyl-2-oxazoline), M.W. 50,000

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 042396.22
also known as 042396-22
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.40
Save 5.75 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.40
Save 5.75 (14%)
Each
Chemical Identifiers
CAS25805-17-8
Molecular Formula(C5H9NO)n
SMILESCCC(=O)N(-*)CC-*
MDL NumberMFCD00134377
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying2%, max ( 1g sample, 2hours @ 125 deg C)
Appearance (Color)Off-white to pale yellow
FormGranular solid
Viscosity5 to 8 cSt ( Kinematic viscosity, 10% aq. solution at 100 degree F)
Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only